Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206609.1 | NP_001193538.1 | 2 | 1728 | ||
NM_003006.4 | NP_002997.2 | 2 | 1663 | |||
GenBank mRNA | AK098315.1 | - | 4 | 1582 | ||
AK127638.1 | - | 3 | 1614 | |||
AK225644.1 | - | 2 | 1559 | |||
AK290395.1 | - | 3 | 1600 | |||
AK295238.1 | - | 3 | 1445 | |||
BC029782.1 | - | 3 | 1612 | 5180824 | ||
U02297.1 | - | 3 | 1532 |