Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206609.1 | NP_001193538.1 | 2 | 1944 | ||
NM_003006.4 | NP_002997.2 | 2 | 1879 | |||
GenBank mRNA | AK098315.1 | - | 4 | 1798 | ||
AK127638.1 | - | 3 | 1830 | |||
AK225644.1 | - | 2 | 1775 | |||
AK295238.1 | - | 3 | 1661 | |||
AK298738.1 | - | 3 | 1555 | |||
BC029782.1 | - | 3 | 1828 | 5180824 |