Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003042.3 | NP_003033.3 | 16 | 2663 | ||
XM_005265410.4 | XP_005265467.1 | 16 | 4020 | |||
XM_005265411.4 | XP_005265468.1 | 16 | 3959 | |||
XM_006713306.3 | XP_006713369.1 | 16 | 2618 | |||
XM_011534025.2 | XP_011532327.1 | 16 | 4710 | |||
XM_011534027.2 | XP_011532329.1 | 16 | 4649 | |||
XM_017007071.1 | XP_016862560.1 | 17 | 4442 | |||
XM_017007072.1 | XP_016862561.1 | 17 | 3752 | |||
GenBank mRNA | AK090958.1 | - | 1 | 164 | ||
BC033904.1 | - | 16 | 2631 | 5264672 | ||
BC057291.1 | - | 16 | 2642 |