Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166496.1 | NP_001159968.1 | 4 | 1624 | ||
NM_003051.3 | NP_003042.3 | 4 | 1155 | |||
XM_011542026.2 | XP_011540328.1 | 4 | 888 | |||
XM_011542027.2 | XP_011540329.1 | 4 | 898 | |||
GenBank mRNA | AK000641.1 | - | 1 | 40 | ||
AK094064.1 | - | 4 | 1624 | |||
AK296688.1 | - | 4 | 938 | |||
AK303603.1 | - | 4 | 997 | |||
AK312500.1 | - | 4 | 998 | |||
AL162079.1 | - | 4 | 985 | |||
BC026317.1 | - | 4 | 998 | 4811581 | ||
BC045664.1 | - | 5 | 1709 | 5298234 | ||
L31801.1 | - | 3 | 804 |