Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135691.2 | NP_001129163.1 | 17 | 2369 | ||
NM_001142324.1 | NP_001135796.1 | 16 | 2273 | |||
NM_001142325.1 | NP_001135797.1 | 16 | 2235 | |||
NM_003053.3 | NP_003044.1 | 16 | 2165 | |||
XM_011544625.1 | XP_011542927.1 | 15 | 2014 | |||
XM_011544626.1 | XP_011542928.1 | 15 | 2002 | |||
XM_011544627.1 | XP_011542929.1 | 15 | 1964 | |||
GenBank mRNA | BC006317.2 | - | 16 | 2182 | 4125784 | |
BC009387.2 | - | 17 | 2186 | 4127678 | ||
U39905.1 | - | 16 | 2165 |