Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 13 | 2233 | ||
NM_001289397.1 | NP_001276326.1 | 13 | 2246 | |||
NM_003070.4 | NP_003061.3 | 13 | 2246 | |||
NM_139045.3 | NP_620614.2 | 13 | 2246 | |||
GenBank mRNA | AK296373.1 | - | 13 | 1634 | ||
X72889.1 | - | 13 | 2234 |