Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 34 | 5423 | ||
NM_001289397.1 | NP_001276326.1 | 33 | 5208 | |||
NM_001289398.1 | NP_001276327.1 | 7 | 1390 | |||
NM_001289399.1 | NP_001276328.1 | 8 | 1377 | |||
NM_001289400.1 | NP_001276329.1 | 8 | 1798 | |||
NM_003070.4 | NP_003061.3 | 34 | 5436 | |||
NM_139045.3 | NP_620614.2 | 33 | 5382 | |||
GenBank mRNA | AK094076.1 | - | 8 | 1798 | ||
AK298045.1 | - | 8 | 1377 | |||
X72889.1 | - | 34 | 5424 |