Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243998.1 | NP_001230927.1 | 5 | 1061 | ||
NM_001243999.1 | NP_001230928.1 | 6 | 1272 | |||
NM_001244000.1 | NP_001230929.1 | 6 | 1241 | |||
NM_003121.4 | NP_003112.2 | 6 | 1276 | |||
GenBank mRNA | AK225225.1 | - | 6 | 1274 | ||
AK225720.1 | - | 6 | 1276 | |||
BC007921.2 | - | 6 | 1255 | 4309499 | ||
BQ064401.1 | - | 1 | 524 | |||
X66079.1 | - | 6 | 1256 | |||
X96998.1 | - | 6 | 1252 |