Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003124.4 | NP_003115.1 | 3 | 799 | ||
GenBank mRNA | AK222942.1 | - | 3 | 787 | ||
AK291856.1 | - | 3 | 759 | |||
BC017310.2 | - | 3 | 763 | 5089862 | ||
M76231.1 | - | 3 | 771 | |||
S71376.1 | - | 1 | 130 |