Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003124.4 | NP_003115.1 | 3 | 1076 | ||
GenBank mRNA | AK222942.1 | - | 3 | 1064 | ||
BC017310.2 | - | 3 | 1040 | 5089862 |