Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003129.3 | NP_003120.2 | 11 | 2922 | ||
XM_011517246.2 | XP_011515548.1 | 11 | 2020 | |||
GenBank mRNA | AF098865.1 | - | 11 | 2151 | ||
AK225085.1 | - | 11 | 2212 | |||
AK301792.1 | - | 11 | 1814 | |||
BC017033.1 | - | 11 | 2154 | 3846346 | ||
BX647400.1 | - | 11 | 2856 | |||
BX647605.1 | - | 11 | 2894 | |||
D78130.1 | - | 11 | 2259 |