Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130440.1 | NP_001123912.1 | 4 | 1376 | ||
NM_003133.5 | NP_003124.1 | 3 | 1252 | |||
GenBank mRNA | AF070649.1 | - | 3 | 1231 | ||
AL050363.1 | - | 1 | 1181 | |||
BC008443.1 | - | 3 | 1236 | 4102222 | ||
BC015094.1 | - | 1 | 1223 | 3930047 | ||
BC017414.1 | - | 1 | 1003 | |||
BC021995.1 | - | 4 | 1356 | 4101294 | ||
BC022415.1 | - | 1 | 1218 | 4243562 | ||
BC064351.1 | - | 3 | 2468 | 4452218 | ||
BC066957.1 | - | 3 | 1239 | 5263635 | ||
BC067845.1 | - | 4 | 1311 | 5287027 | ||
BC106005.1 | - | 1 | 1209 | 6026221 | ||
EF488978.1 | - | 4 | 1360 | |||
U20998.1 | - | 3 | 1229 |