Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114121.2 | NP_001107593.1 | 3 | 1173 | ||
NM_001114122.2 | NP_001107594.1 | 3 | 1173 | |||
NM_001244846.1 | NP_001231775.1 | 3 | 1173 | |||
NM_001274.5 | NP_001265.2 | 3 | 516 | |||
NR_045205.1 | 3 | 710 | ||||
XM_017017146.1 | XP_016872635.1 | 3 | 360 | |||
GenBank mRNA | AB451222.1 | - | 2 | 276 | ||
AB451345.1 | - | 2 | 276 | |||
AF016582.1 | - | 2 | 310 | |||
AF032874.1 | - | 2 | 276 | |||
AK292549.1 | - | 3 | 710 | |||
AK293143.1 | - | 3 | 338 | |||
BC004202.2 | - | 3 | 335 | 3530606 | ||
BC017575.1 | - | 3 | 434 | 4091997 | ||
BP207812.1 | - | 4 | 478 | |||
DA567616.1 | - | 3 | 407 |