Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 22 | 3109 | ||
NM_001178079.1 | NP_001171550.1 | 22 | 3035 | |||
NM_001178080.1 | NP_001171551.1 | 21 | 2953 | |||
NM_001178081.1 | NP_001171552.1 | 20 | 2814 | |||
NM_003153.4 | NP_003144.3 | 22 | 3090 | |||
NR_033659.1 | 21 | 2951 | ||||
XM_006719575.2 | XP_006719638.1 | 23 | 3087 | |||
XM_011538703.2 | XP_011537005.1 | 23 | 3164 | |||
XM_011538704.2 | XP_011537006.1 | 24 | 3175 | |||
XM_011538705.2 | XP_011537007.1 | 24 | 3285 | |||
XM_011538706.2 | XP_011537008.1 | 24 | 3141 | |||
XM_011538707.2 | XP_011537009.1 | 23 | 3126 | |||
XM_011538708.2 | XP_011537010.1 | 22 | 2968 | |||
XM_011538709.2 | XP_011537011.1 | 23 | 3002 | |||
GenBank mRNA | AK291044.1 | - | 22 | 3036 | ||
AK298721.1 | - | 20 | 2634 | |||
AK316236.1 | - | 22 | 2510 | |||
AY615283.1 | - | 8 | 1668 | |||
BC075852.1 | - | 22 | 2891 | 5530399 | ||
U16031.1 | - | 22 | 2949 |