Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003168.2 | NP_003159.1 | 5 | 683 | ||
NR_073470.1 | 5 | 685 | ||||
GenBank mRNA | BC002802.2 | - | 5 | 641 | 3636217 | |
BX331655.2 | - | 5 | 647 | |||
U38817.1 | - | 5 | 622 | |||
U38818.1 | - | 5 | 641 | |||
U43923.1 | - | 5 | 647 |