Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280787.1 | NP_001267716.1 | 7 | 769 | ||
NM_003172.3 | NP_003163.1 | 8 | 821 | |||
XM_011518942.1 | XP_011517244.1 | 7 | 871 | |||
GenBank mRNA | AK291122.1 | - | 8 | 804 | ||
BC028314.1 | - | 8 | 809 | 5190921 | ||
BC071658.1 | - | 8 | 826 | 5539506 | ||
KU178341.1 | - | 8 | 789 | |||
KU178342.1 | - | 7 | 737 | |||
KU178343.1 | - | 6 | 603 | |||
Z35093.1 | - | 8 | 803 |