Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 17 | 1988 | ||
NM_001313965.1 | NP_001300894.1 | 16 | 1786 | |||
NM_001313966.1 | NP_001300895.1 | 15 | 1590 | |||
NM_003234.3 | NP_003225.2 | 17 | 2129 | |||
GenBank mRNA | AB209254.1 | - | 17 | 1892 | ||
AK291723.1 | - | 17 | 2129 | |||
AK294758.1 | - | 15 | 1520 | |||
BC001188.1 | - | 17 | 1902 | 3354176 | ||
M11507.1 | - | 17 | 1941 | |||
X01060.1 | - | 17 | 2109 |