Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 19 | 3028 | ||
NM_001313965.1 | NP_001300894.1 | 18 | 2826 | |||
NM_001313966.1 | NP_001300895.1 | 17 | 2630 | |||
NM_003234.3 | NP_003225.2 | 19 | 3169 | |||
GenBank mRNA | BC001188.1 | - | 19 | 2942 | 3354176 | |
BX537966.1 | - | 18 | 2756 | |||
X01060.1 | - | 19 | 3149 |