Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003268.5 | NP_003259.2 | 6 | 2982 | ||
XM_005273241.4 | XP_005273298.2 | 6 | 2928 | |||
XM_005273242.4 | XP_005273299.2 | 7 | 3048 | |||
XM_005273243.4 | XP_005273300.2 | 5 | 2834 | |||
XM_006711504.3 | XP_006711567.1 | 5 | 2873 | |||
XM_006711505.3 | XP_006711568.1 | 4 | 2779 | |||
XM_006711506.3 | XP_006711569.1 | 3 | 2581 | |||
XM_011509937.2 | XP_011508239.1 | 6 | 2989 | |||
XM_017002208.1 | XP_016857697.1 | 5 | 2895 | |||
GenBank mRNA | AB060695.1 | - | 6 | 3044 | ||
AB445645.1 | - | 1 | 2341 | |||
AF051151.1 | - | 6 | 3044 | |||
BC109118.1 | - | 4 | 2679 | 40035617 | ||
BC109119.1 | - | 4 | 2679 | 40035618 | ||
U88881.1 | - | 1 | 862 |