Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093771.2 | NP_001087240.1 | 17 | 3637 | ||
NM_001261445.1 | NP_001248374.1 | 15 | 4059 | |||
NM_001261446.1 | NP_001248375.1 | 14 | 3614 | |||
NM_003330.3 | NP_003321.3 | 15 | 3974 | |||
NM_182729.2 | NP_877393.1 | 15 | 3737 | |||
NM_182742.2 | NP_877419.1 | 14 | 3864 | |||
NM_182743.2 | NP_877420.1 | 14 | 3627 | |||
GenBank mRNA | AF077367.1 | - | 14 | 3290 | ||
AJ001050.1 | - | 13 | 3200 | |||
AK128515.1 | - | 15 | 3659 | |||
AK129977.1 | - | 6 | 2307 | |||
BC018122.1 | - | 15 | 3350 | 3883490 | ||
D88687.1 | - | 15 | 3416 | |||
S79851.1 | - | 15 | 3601 | |||
X91247.1 | - | 15 | 3601 |