Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018056.2 | NP_001018066.1 | 5 | 930 | ||
NM_001322225.1 | NP_001309154.1 | 4 | 807 | |||
NM_001322226.1 | NP_001309155.1 | 4 | 807 | |||
NM_003383.4 | NP_003374.3 | 5 | 930 | |||
GenBank mRNA | AB208822.1 | - | 6 | 686 | ||
AK095744.1 | - | 17 | ||||
BC136562.1 | - | 5 | 651 | 9020552 | ||
BC142653.1 | - | 5 | 592 | 40148037 | ||
BC144245.1 | - | 5 | 651 | 9052765 | ||
D16493.1 | - | 5 | 584 | |||
D16494.1 | - | 5 | 515 | |||
L20470.1 | - | 5 | 905 | |||
L22431.1 | - | 5 | 627 |