Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 16 | 2479 | ||
XM_005264544.1 | XP_005264601.1 | 15 | 2456 | |||
XM_005264546.1 | XP_005264603.1 | 14 | 1587 | |||
XM_006712094.2 | XP_006712157.1 | 17 | 2622 | |||
XM_011533097.1 | XP_011531399.1 | 16 | 1788 | |||
XM_011533098.1 | XP_011531400.1 | 16 | 2456 | |||
XM_011533099.2 | XP_011531401.1 | 16 | 3429 | |||
GenBank mRNA | AK124769.1 | - | 5 | 1356 | ||
AK289920.1 | - | 16 | 2158 | |||
AK299039.1 | - | 7 | 881 | |||
AK312426.1 | - | 16 | 1916 | |||
AK316481.1 | - | 7 | 895 | |||
AL833550.1 | - | 15 | 4424 | |||
BC032847.2 | - | 17 | 1880 | 5267242 | ||
BX647758.1 | - | 16 | 2461 | |||
CR749840.1 | - | 17 | 2253 | |||
D89729.1 | - | 15 | 1769 | |||
Y08614.2 | - | 16 | 1789 |