Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 16, 17 | 2623 | ||
XM_005264544.1 | XP_005264601.1 | 15, 16 | 2600 | |||
XM_005264546.1 | XP_005264603.1 | 14, 15 | 1731 | |||
XM_006712094.2 | XP_006712157.1 | 17, 18 | 2766 | |||
XM_011533097.1 | XP_011531399.1 | 16, 17 | 1932 | |||
XM_011533099.2 | XP_011531401.1 | 16, 17 | 3573 | |||
GenBank mRNA | AK124769.1 | - | 5, 6 | 1500 | ||
AK289920.1 | - | 16, 17 | 2302 | |||
AK299039.1 | - | 7, 8 | 1025 | |||
AK312426.1 | - | 16, 17 | 2060 | |||
AK316481.1 | - | 7, 8 | 1039 | |||
AL833550.1 | - | 15, 16 | 4568 | |||
BC032847.2 | - | 17, 18 | 2024 | 5267242 | ||
BX647758.1 | - | 16, 17 | 2605 | |||
CR749840.1 | - | 17, 18 | 2397 | |||
D89729.1 | - | 15, 16 | 1913 | |||
Y08614.2 | - | 16, 17 | 1933 |