Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286055.1 | NP_001272984.1 | 6 | 1175 | ||
NM_001286056.1 | NP_001272985.1 | 7 | 1103 | |||
NM_001839.4 | NP_001830.1 | 7 | 1313 | |||
XM_017000245.1 | XP_016855734.1 | 9 | 3725 | |||
GenBank mRNA | AK294141.1 | - | 7 | 1064 | ||
AK298145.1 | - | 6 | 1095 | |||
AK316398.1 | - | 7 | 1079 | |||
BC025372.2 | - | 7 | 1197 | 3505668 | ||
S80562.1 | - | 7 | 1067 |