Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145446.1 | NP_001138918.1 | 3 | 732 | ||
NM_001145447.1 | NP_001138919.1 | 3 | 937 | |||
NM_001145448.1 | NP_001138920.1 | 3 | 856 | |||
NM_003454.3 | NP_003445.2 | 3 | 937 | |||
NM_198087.2 | NP_932353.1 | 3 | 732 | |||
NM_198088.2 | NP_932354.1 | 3 | 856 | |||
XM_005255556.1 | XP_005255613.1 | 4 | 889 | |||
XM_006720940.2 | XP_006721003.1 | 3 | 1260 | |||
XM_006720941.2 | XP_006721004.1 | 3 | 732 | |||
XM_017023647.1 | XP_016879136.1 | 4 | 1028 | |||
XM_017023648.1 | XP_016879137.1 | 3 | 588 | |||
GenBank mRNA | AF060866.1 | - | 3 | 541 | ||
AK056010.1 | - | 3 | 937 | |||
AK315036.1 | - | 3 | 470 | |||
BC012909.1 | - | 3 | 918 | 4470685 | ||
BC025293.1 | - | 3 | 916 | |||
BC032575.1 | - | 3 | 855 | 5490702 | ||
BC054005.1 | - | 3 | 686 | 6166814 | ||
BI459882.1 | - | 3 | 562 | |||
BU177850.1 | - | 3 | 684 | |||
Y14443.1 | - | 3 | 635 |