Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145446.1 | NP_001138918.1 | 4 | 816 | ||
NM_001145447.1 | NP_001138919.1 | 4 | 1021 | |||
NM_001145448.1 | NP_001138920.1 | 4 | 940 | |||
NM_003454.3 | NP_003445.2 | 4 | 1021 | |||
NM_198087.2 | NP_932353.1 | 4 | 813 | |||
NM_198088.2 | NP_932354.1 | 4 | 940 | |||
XM_005255556.1 | XP_005255613.1 | 5 | 973 | |||
XM_006720940.2 | XP_006721003.1 | 4 | 1344 | |||
XM_006720941.2 | XP_006721004.1 | 4 | 816 | |||
XM_017023647.1 | XP_016879136.1 | 5 | 1112 | |||
XM_017023648.1 | XP_016879137.1 | 4 | 672 | |||
GenBank mRNA | AF060866.1 | - | 4 | 625 | ||
AK056010.1 | - | 4 | 1021 | |||
AK315036.1 | - | 4 | 554 | |||
BC012909.1 | - | 4 | 1002 | 4470685 | ||
BC025293.1 | - | 4 | 1000 | |||
BC032575.1 | - | 4 | 939 | 5490702 | ||
BC054005.1 | - | 4 | 770 | 6166814 | ||
BI459882.1 | - | 4 | 646 | |||
Y14443.1 | - | 4 | 716 |