Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 10 | 1384 | ||
XM_005274606.3 | XP_005274663.1 | 11 | 1526 | |||
XM_011545592.2 | XP_011543894.1 | 11 | 1355 | |||
XM_011545594.2 | XP_011543896.1 | 9 | 805 | |||
XM_011545595.1 | XP_011543897.1 | 9 | 815 | |||
XM_011545597.1 | XP_011543899.1 | 11 | 1440 | |||
XM_011545598.1 | XP_011543900.1 | 1 | 49 | |||
XM_017029909.1 | XP_016885398.1 | 10 | 960 | |||
XM_017029910.1 | XP_016885399.1 | 11 | 1168 | |||
XM_017029911.1 | XP_016885400.1 | 1 | 78 | |||
GenBank mRNA | AK289677.1 | - | 10 | 1365 | ||
AK290354.1 | - | 10 | 1369 | |||
GU727634.1 | - | 9 | 1025 | |||
Y15164.1 | - | 10 | 1336 | |||
Y16355.1 | - | 10 | 1336 |