Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270942.1 | NP_001257871.1 | 3 | 459 | ||
NM_001270943.1 | NP_001257872.1 | 2 | 226 | |||
NM_001270944.1 | NP_001257873.1 | 2 | 190 | |||
NM_003709.3 | NP_003700.1 | 2 | 669 | |||
XM_005246926.3 | XP_005246983.1 | 3 | 455 | |||
XM_006712815.3 | XP_006712878.1 | 3 | 459 | |||
XM_011512072.2 | XP_011510374.1 | 3 | 633 | |||
XM_011512073.2 | XP_011510375.1 | 4 | 641 | |||
XM_011512074.2 | XP_011510376.1 | 3 | 236 | |||
XM_011512075.1 | XP_011510377.1 | 2 | 642 | |||
XM_017005161.1 | XP_016860650.1 | 3 | 455 | |||
XM_017005162.1 | XP_016860651.1 | 2 | 205 | |||
XM_017005163.1 | XP_016860652.1 | 2 | 642 | |||
XM_017005164.1 | XP_016860653.1 | 2 | 654 | |||
GenBank mRNA | AB015132.1 | - | 2 | 544 | ||
AK027371.1 | - | 2 | 370 | |||
AK297312.1 | - | 2 | 177 | |||
AK314439.1 | - | 2 | 600 | |||
BC012919.1 | - | 3 | 450 | 4509200 | ||
BX537890.1 | - | 2 | 226 | |||
CR542112.1 | - | 2 | 221 | |||
DA800775.1 | - | 3 | 459 | |||
DC370575.1 | - | 2 | 190 |