Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003725.3 | NP_003716.2 | 5 | 981 | ||
XM_005269207.1 | XP_005269264.1 | 6 | 1130 | |||
XM_005269208.1 | XP_005269265.1 | 6 | 1134 | |||
XM_005269209.1 | XP_005269266.1 | 6 | 1121 | |||
XM_006719672.1 | XP_006719735.1 | 6 | 1129 | |||
XM_011538925.1 | XP_011537227.1 | 6 | 1124 | |||
XM_011538926.1 | XP_011537228.1 | 6 | 1114 | |||
XM_011538927.1 | XP_011537229.1 | 6 | 1105 | |||
GenBank mRNA | AF016509.1 | - | 4 | 765 | ||
AF223225.1 | - | 5 | 852 | |||
AK313423.1 | - | 5 | 891 | |||
BC020710.1 | - | 5 | 875 | 4700726 | ||
U89281.1 | - | 5 | 870 |