Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003786.3 | NP_003777.2 | 23 | 3310 | ||
XM_005257763.3 | XP_005257820.1 | 22 | 3132 | |||
XM_011525422.2 | XP_011523724.1 | 22 | 3237 | |||
XM_011525423.1 | XP_011523725.1 | 23 | 3429 | |||
XM_011525424.2 | XP_011523726.1 | 18 | 2660 | |||
XM_011525425.2 | XP_011523727.1 | 22 | 3137 | |||
XM_017025265.1 | XP_016880754.1 | 19 | 3173 | |||
XM_017025266.1 | XP_016880755.1 | 23 | 3429 | |||
GenBank mRNA | AB010887.1 | - | 23 | 3244 | ||
AB208954.1 | - | 23 | 3250 | |||
AF009670.1 | - | 23 | 3303 | |||
AF083552.1 | - | 23 | 3247 | |||
AF085690.1 | - | 23 | 3266 | |||
AF085691.1 | - | 23 | 3266 | |||
AF085692.1 | - | 23 | 3321 | |||
AF104943.1 | - | 23 | 3234 | |||
BC137347.1 | - | 23 | 3261 | 9021345 | ||
BC137348.1 | - | 23 | 3261 | 9021346 | ||
U83659.1 | - | 1 | 106 | |||
Y17151.2 | - | 23 | 3272 |