Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 22 | 3279 | ||
XM_011540326.1 | XP_011538628.1 | 22 | 3378 | |||
XM_011540327.2 | XP_011538629.1 | 19 | 3044 | |||
XM_011540328.2 | XP_011538630.1 | 19 | 2872 | |||
XM_011540329.2 | XP_011538631.1 | 22 | 3489 | |||
XM_017016877.1 | XP_016872366.1 | 21 | 3341 | |||
XM_017016878.1 | XP_016872367.1 | 21 | 3171 | |||
XM_017016879.1 | XP_016872368.1 | 21 | 3295 | |||
XM_017016880.1 | XP_016872369.1 | 21 | 3299 | |||
XM_017016881.1 | XP_016872370.1 | 20 | 3148 | |||
GenBank mRNA | AB208843.1 | - | 9 | 1568 | ||
AF038362.1 | - | 22 | 3298 | |||
AJ001017.2 | - | 22 | 3193 | |||
BC112201.1 | - | 22 | 3263 | 8327669 |