Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165924.1 | NP_001159396.1 | 2 | 2353 | ||
NM_004098.3 | NP_004089.1 | 3 | 2538 | |||
GenBank mRNA | AF301598.1 | - | 3 | 2538 | ||
AK055041.1 | - | 3 | 1939 | |||
AL161811.1 | - | 3 | 2041 |