Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 9, 10 | 1223 | ||
GenBank mRNA | AK124421.1 | - | 2, 3 | 1651 | ||
AK291493.1 | - | 9, 10 | 1100 | |||
BC005279.1 | - | 9, 10 | 1095 | 3949850 | ||
BT007313.1 | - | 9, 10 | 1073 | |||
X67318.1 | - | 9, 10 | 1080 |