Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004135.3 | NP_004126.1 | 5, 6 | 550 | ||
NM_174869.2 | NP_777358.1 | 5, 6 | 550 | |||
GenBank mRNA | BC000933.2 | - | 5, 6 | 396 | 3447003 | |
BC001902.1 | - | 5, 6 | 391 | 3543839 | ||
BI831096.1 | - | 5, 6 | 548 | |||
U40272.1 | - | 5, 6 | 545 | |||
U69268.1 | - | 1, 2 | 101 | |||
Z68907.1 | - | 5, 6 | 432 |