Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318369.1 | NP_001305298.1 | 11, 12 | 2011 | ||
NM_004211.4 | NP_004202.3 | 12, 13 | 2150 | |||
XM_017018544.1 | XP_016874033.1 | 8, 9 | 1081 | |||
XM_017018545.1 | XP_016874034.1 | 8, 9 | 1074 | |||
GenBank mRNA | AF085412.1 | - | 12, 13 | 2158 | ||
AF117999.1 | - | 11, 12 | 1877 | |||
AF142501.1 | - | 11, 12 | 1895 | |||
AF352733.1 | - | 12, 13 | 1877 | |||
BC096319.3 | - | 12, 13 | 1956 | 40007841 | ||
BC096320.3 | - | 12, 13 | 1956 | 40007842 | ||
BC096321.1 | - | 12, 13 | 1956 | 40007845 | ||
BC096322.3 | - | 11, 12 | 1817 |