Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314052.1 | NP_001300981.1 | 4 | 2396 | ||
NM_004235.5 | NP_004226.3 | 5 | 2294 | |||
GenBank mRNA | AF105036.1 | - | 5 | 2048 | ||
AK095134.1 | - | 3 | 1800 | |||
AK095894.1 | - | 1 | 1543 | |||
BC030811.1 | - | 3 | 1850 | 4703002 |