Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004244.5 | NP_004235.4 | 4 | 785 | ||
NM_203416.3 | NP_981961.2 | 4 | 785 | |||
XM_005253528.4 | XP_005253585.1 | 4 | 775 | |||
XM_005253529.4 | XP_005253586.1 | 4 | 775 | |||
XM_017020231.1 | XP_016875720.1 | 4 | 775 | |||
GenBank mRNA | AK226017.1 | - | 4 | 735 | ||
BC051281.1 | - | 4 | 717 | 6280265 | ||
DQ058615.1 | - | 4 | 657 | |||
Z22968.1 | - | 4 | 743 | |||
Z22969.1 | - | 4 | 743 | |||
Z22970.1 | - | 4 | 743 | |||
Z22971.1 | - | 4 | 743 |