Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 3 | 523 | ||
NM_001184733.1 | NP_001171662.1 | 3 | 271 | |||
NM_001184736.1 | NP_001171665.1 | 2 | 186 | |||
NM_004254.3 | NP_004245.2 | 3 | 544 | |||
XM_011545364.1 | XP_011543666.1 | 2 | 119 | |||
GenBank mRNA | AB042505.1 | - | 3 | 526 | ||
AF097491.1 | - | 2 | 402 | |||
AK298341.1 | - | 3 | 241 | |||
AK298351.1 | - | 4 | 496 | |||
AK307850.1 | - | 2 | 151 | |||
AK313187.1 | - | 3 | 517 | |||
BC022387.1 | - | 3 | 538 | 4608542 | ||
BP276255.1 | - | 3 | 241 | |||
DA633836.1 | - | 3 | 493 |