Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 5 | 712 | ||
NM_001281502.1 | NP_001268431.1 | 5 | 781 | |||
NM_004265.3 | NP_004256.1 | 5 | 1053 | |||
GenBank mRNA | AF084559.1 | - | 5 | 845 | ||
AF108658.1 | - | 5 | 638 | |||
AF126799.1 | - | 5 | 708 | |||
AK074925.1 | - | 3 | 260 | |||
AK074939.1 | - | 5 | 785 | |||
AK074991.1 | - | 5 | 521 | |||
AK290291.1 | - | 5 | 659 | |||
AK299762.1 | - | 5 | 781 | |||
AL050118.1 | - | 3 | 297 | |||
BC009011.1 | - | 5 | 787 | 4181037 |