Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 12 | 1937 | ||
NM_001281502.1 | NP_001268431.1 | 12 | 2006 | |||
NM_004265.3 | NP_004256.1 | 12 | 2278 | |||
GenBank mRNA | AF084559.1 | - | 12 | 2070 | ||
AF108658.1 | - | 12 | 1863 | |||
AF126799.1 | - | 12 | 1933 | |||
AK027459.1 | - | 6 | 1073 | |||
AK027513.1 | - | 6 | 1092 | |||
AK027577.1 | - | 6 | 1117 | |||
AK074925.1 | - | 10 | 1485 | |||
AK074939.1 | - | 12 | 2010 | |||
AK074991.1 | - | 12 | 1746 | |||
AK123240.1 | - | 7 | 1907 | |||
AK290291.1 | - | 12 | 1884 | |||
AL050118.1 | - | 10 | 1522 |