Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 12 | 2453 | ||
NM_001281502.1 | NP_001268431.1 | 12 | 2522 | |||
NM_004265.3 | NP_004256.1 | 12 | 2794 | |||
GenBank mRNA | AF084559.1 | - | 12 | 2586 | ||
AF108658.1 | - | 12 | 2379 | |||
AF126799.1 | - | 12 | 2449 | |||
AK027459.1 | - | 6 | 1589 | |||
AK027513.1 | - | 6 | 1608 | |||
AK027577.1 | - | 6 | 1633 | |||
AK074925.1 | - | 10 | 2001 | |||
AK074939.1 | - | 12 | 2526 | |||
AK074991.1 | - | 12 | 2262 | |||
AK123240.1 | - | 7 | 2423 | |||
AL050118.1 | - | 10 | 2038 |