Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079846.1 | NP_001073315.1 | 2 | 827 | ||
NM_004380.2 | NP_004371.2 | 2 | 827 | |||
XM_005255124.4 | XP_005255181.1 | 2 | 1434 | |||
XM_005255125.4 | XP_005255182.1 | 2 | 1434 | |||
XM_006720848.3 | XP_006720911.1 | 2 | 1434 | |||
XM_011522382.2 | XP_011520684.1 | 2 | 1434 | |||
XM_017022944.1 | XP_016878433.1 | 2 | 1434 | |||
GenBank mRNA | AB210043.1 | - | 2 | 827 | ||
U47741.1 | - | 3 | 821 | |||
U85962.3 | - | 2 | 1441 |