Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079846.1 | NP_001073315.1 | 7 | 1864 | ||
NM_004380.2 | NP_004371.2 | 8 | 1978 | |||
XM_005255124.4 | XP_005255181.1 | 8 | 2585 | |||
XM_005255125.4 | XP_005255182.1 | 8 | 2585 | |||
XM_006720848.3 | XP_006720911.1 | 8 | 2585 | |||
XM_011522381.2 | XP_011520683.1 | 7 | 1467 | |||
XM_011522382.2 | XP_011520684.1 | 8 | 2585 | |||
XM_017022944.1 | XP_016878433.1 | 8 | 2585 | |||
GenBank mRNA | AB210043.1 | - | 7 | 1864 | ||
AK304771.1 | - | 5 | 752 | |||
U47741.1 | - | 9 | 1972 | |||
U85962.3 | - | 8 | 2592 |