Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 2996 | ||
XM_005253334.2 | XP_005253391.1 | 14 | 2996 | |||
XM_011520599.2 | XP_011518901.1 | 14 | 2999 | |||
XM_011520600.2 | XP_011518902.1 | 13 | 2873 | |||
XM_011520603.2 | XP_011518905.1 | 14 | 3103 | |||
XM_017018955.1 | XP_016874444.1 | 14 | 3391 | |||
XM_017018956.1 | XP_016874445.1 | 13 | 2849 | |||
XM_017018957.1 | XP_016874446.1 | 13 | 2822 | |||
XM_017018958.1 | XP_016874447.1 | 13 | 2798 | |||
GenBank mRNA | AB209063.1 | - | 15 | 3172 | ||
BC002871.2 | - | 7 | 1272 | |||
BC017748.1 | - | 7 | 1333 | |||
BC073964.1 | - | 3 | 2844 | 5788132 | ||
BC157850.1 | - | 2 | 2902 | 9057069 | ||
BX640639.1 | - | 15 | 2603 | |||
U89277.1 | - | 14 | 2975 |