Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 3777 | ||
XM_005253334.2 | XP_005253391.1 | 15 | 3777 | |||
XM_011520599.2 | XP_011518901.1 | 15 | 3780 | |||
XM_011520600.2 | XP_011518902.1 | 14 | 3654 | |||
XM_011520603.2 | XP_011518905.1 | 15 | 3884 | |||
XM_017018955.1 | XP_016874444.1 | 15 | 4172 | |||
XM_017018956.1 | XP_016874445.1 | 14 | 3630 | |||
XM_017018957.1 | XP_016874446.1 | 14 | 3603 | |||
XM_017018958.1 | XP_016874447.1 | 14 | 3579 | |||
GenBank mRNA | AB209063.1 | - | 16 | 3953 | ||
BC002871.2 | - | 8 | 2053 | |||
BC017748.1 | - | 8 | 2114 | |||
BC073964.1 | - | 3 | 3625 | 5788132 |