Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199880.1 | NP_001186809.1 | 2 | 3133 | ||
NM_001199881.1 | NP_001186810.1 | 2 | 3127 | |||
NM_004430.2 | NP_004421.2 | 2 | 3557 | |||
XM_005273425.3 | XP_005273482.1 | 2 | 3508 | |||
XM_005273426.3 | XP_005273483.1 | 3 | 3181 | |||
XM_011544429.2 | XP_011542731.1 | 3 | 3293 | |||
GenBank mRNA | S40832.1 | - | 2 | 3556 | ||
X63741.1 | - | 2 | 3551 |