Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EZH2 SIRNA Assays ›
Gene Symbol:
EZH2
Gene Name:
enhancer of zeste 2 polycomb repressive complex 2 subunit
Gene Aliases:
ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
Chromosome Location:
Chr.7: 148807372 - 148884662 on Build GRCh38
UniGene ID:
Hs.444082
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107418
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EZH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001203247.1
NP_001190176.1
10
1267
NM_001203248.1
NP_001190177.1
10
1240
NM_001203249.1
NP_001190178.1
10
1367
NM_004456.4
NP_004447.2
10
1282
NM_152998.2
NP_694543.1
9
1150
XM_005249962.4
XP_005250019.1
10
3708
XM_005249963.4
XP_005250020.1
10
3681
XM_005249964.4
XP_005250021.1
10
3682
XM_011515883.2
XP_011514185.1
11
1257
XM_011515884.2
XP_011514186.1
10
1389
XM_011515885.2
XP_011514187.1
10
3698
XM_011515886.2
XP_011514188.1
11
1406
XM_011515887.2
XP_011514189.1
10
1255
XM_011515888.2
XP_011514190.1
10
1377
XM_011515889.2
XP_011514191.1
10
1179
XM_011515890.2
XP_011514192.1
9
3608
XM_011515891.2
XP_011514193.1
8
1342
XM_011515892.2
XP_011514194.1
10
3726
XM_011515893.2
XP_011514195.1
9
3591
XM_011515894.2
XP_011514196.1
9
1107
XM_011515895.2
XP_011514197.1
9
3581
XM_011515896.2
XP_011514198.1
9
3592
XM_011515897.2
XP_011514199.1
11
1433
XM_011515898.2
XP_011514200.1
11
1360
XM_011515899.2
XP_011514201.1
10
3728
XM_011515901.2
XP_011514203.1
10
3728
XM_017011817.1
XP_016867306.1
10
3724
XM_017011818.1
XP_016867307.1
11
1391
XM_017011819.1
XP_016867308.1
10
3709
XM_017011820.1
XP_016867309.1
9
1138
XM_017011821.1
XP_016867310.1
11
1418
GenBank mRNA
AB208895.1
-
11
1283
AK092676.1
-
9
1049
AK293239.1
-
10
1178
AK302216.1
-
10
1367
AK303585.1
-
10
1205
AK314291.1
-
9
1092
BC010858.2
-
10
1211
3901250
JF327843.1
-
8
957
U61145.1
-
10
1163
X95653.1
-
10
1131
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of epithelial to mesenchymal transition
regulation of gliogenesis
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
cerebellar cortex development
hippocampus development
response to estradiol
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of cell proliferation
regulation of circadian rhythm
positive regulation of MAP kinase activity
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of GTPase activity
negative regulation of epidermal cell differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
negative regulation of retinoic acid receptor signaling pathway
rhythmic process
negative regulation of striated muscle cell differentiation
cellular response to hydrogen peroxide
G1 to G0 transition
histone H3-K27 methylation
protein localization to chromatin
positive regulation of protein serine/threonine kinase activity
positive regulation of dendrite development
negative regulation of G1/S transition of mitotic cell cycle
core promoter binding
DNA binding
chromatin binding
RNA binding
protein binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
chromatin DNA binding
histone methyltransferase activity
ribonucleoprotein complex binding
sequence-specific DNA binding
histone methyltransferase activity (H3-K27 specific)
promoter-specific chromatin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-zz5lq:80/10.239.165.3:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0