Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303422.1 | NP_001290351.1 | 9, 10 | 1140 | ||
NM_004490.2 | NP_004481.2 | 10, 11 | 1769 | |||
XM_017003899.1 | XP_016859388.1 | 10, 11 | 1206 | |||
GenBank mRNA | AK301961.1 | - | 9, 10 | 1085 | ||
AK307744.1 | - | 10, 11 | 1282 | |||
AK313153.1 | - | 10, 11 | 1398 | |||
BC053559.1 | - | 10, 11 | 1239 | 6162863 | ||
L76687.1 | - | 10, 11 | 1768 |