Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004502.3 | NP_004493.3 | 2 | 840 | ||
GenBank mRNA | AK223249.1 | - | 2 | 839 | ||
AK290653.1 | - | 2 | 806 | |||
BC015345.1 | - | 2 | 787 | 4413080 | ||
BQ233410.1 | - | 1 | 158 | |||
M16937.1 | - | 2 | 836 | |||
S49765.1 | - | 2 | 443 |