Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267809.1 | NP_001254738.1 | 13, 14 | 1132 | ||
NM_004515.3 | NP_004506.2 | 13, 14 | 1113 | |||
GenBank mRNA | AK223326.1 | - | 13, 14 | 1033 | ||
AK302214.1 | - | 13, 14 | 1076 | |||
AK313364.1 | - | 13, 14 | 1072 | |||
BC000382.2 | - | 13, 14 | 1038 | 2820505 | ||
HQ322261.1 | - | 12, 13 | 1007 | |||
HQ322262.1 | - | 12, 13 | 1007 | |||
U10323.1 | - | 13, 14 | 1046 |